665193
Lanthanum(III) isopropoxide
98%
Synonym(s):
Lanthanum isopropylate, Lanthanum(III) tri-isopropoxide, Tris(isopropoxy)lanthanum(III)
About This Item
Recommended Products
Assay
98%
form
solid
reaction suitability
core: lanthanum
reagent type: catalyst
SMILES string
CC(C)O[La](OC(C)C)OC(C)C
InChI
1S/3C3H7O.La/c3*1-3(2)4;/h3*3H,1-2H3;/q3*-1;+3
InChI key
SORGMJIXNUWMMR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
accessory
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
Shibasaki's research focuses on novel asymmetric catalytic systems for streamlined synthesis of enantioenriched chiral building blocks.
Shibasaki's research focuses on novel asymmetric catalytic systems for streamlined synthesis of enantioenriched chiral building blocks.
Shibasaki's research focuses on novel asymmetric catalytic systems for streamlined synthesis of enantioenriched chiral building blocks.
Shibasaki's research focuses on novel asymmetric catalytic systems for streamlined synthesis of enantioenriched chiral building blocks.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service