Skip to Content
Merck
All Photos(1)

Documents

659576

Sigma-Aldrich

Glycosyloxyethyl methacrylate solution

5 % (w/v) in ethanol

Synonym(s):

GEMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11O5)OCH2CH2OCOC(=CH2)CH3
Molecular Weight:
292.28
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

concentration

5 % (w/v) in ethanol

refractive index

n20/D 1.366

density

0.808 g/mL at 25 °C

SMILES string

CC(=C)C(=O)OCCOC1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C12H20O8/c1-6(2)11(17)18-3-4-19-12-10(16)9(15)8(14)7(5-13)20-12/h7-10,12-16H,1,3-5H2,2H3/t7-,8-,9+,10-,12?/m1/s1

InChI key

KUQRLZZWFINMDP-BGNLRFAXSA-N

Application

Forms copolymer with butyl acrylate which acts as a pressure-sensitive biomedical adhesive.
Monomer for hydrogels used in drug delivery.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jose Gregorio Soto-Campos et al.
BMC pulmonary medicine, 13, 73-73 (2013-12-11)
There is limited information on the causes of death in asthma patients.To determine the causes of death in hospitalized asthmatic patients and to compare with those observed in COPD patients and non-respiratory individuals, with a particular interest in associations with
Allan S Hoffman
Advanced drug delivery reviews, 54(1), 3-12 (2002-01-05)
This article reviews the composition and synthesis of hydrogels, the character of their absorbed water, and permeation of solutes within their swollen matrices. The most important properties of hydrogels relevant to their biomedical applications are also identified, especially for use
Miyata, T. et al.
Journal of Applied Polymer Science. Applied Polymer Symposium, 56, 1615-1615 (2003)
Sarah D Kowitt et al.
Nicotine & tobacco research : official journal of the Society for Research on Nicotine and Tobacco, 22(6), 958-966 (2019-05-28)
We investigated associations between exposure to tobacco outlets within activity spaces and daily tobacco use, and whether exposure to adults or peers using tobacco mediate these relationships. We used Geographic Ecological Momentary Assessment data over 14 days from 85 youth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service