Skip to Content
Merck
All Photos(2)

Key Documents

659274

Sigma-Aldrich

Potassium allyltrifluoroborate

95%

Synonym(s):

Potassium trifluoro(prop-2-enyl)borate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5BF3K
CAS Number:
Molecular Weight:
147.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

mp

>300 °C

SMILES string

[K+].F[B-](F)(F)CC=C

InChI

1S/C3H5BF3.K/c1-2-3-4(5,6)7;/h2H,1,3H2;/q-1;+1

InChI key

TVPZAMJXLCDMIT-UHFFFAOYSA-N

Related Categories

Application

Organotrifluoroborate involved in:
  • Catalytic allylboration
  • Stereoselective nucleophilic addition
  • Pd-catalyzed heterocyclizations
  • Oxidation reactions and Oxidative Mannich reactions
  • Cross-coupling reactions

Organotrifluoroborates as versatile and stable boronic acid surrogates.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Bench-stable Potassium Organotrifluoroborates enable diverse C-C bond formation reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service