Skip to Content
Merck
All Photos(1)

Key Documents

651494

Sigma-Aldrich

Gallium phosphide

(single crystal substrate), <111>, diam. × thickness 2 in. × 0.5 mm

Synonym(s):

Gallium monophosphide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GaP
CAS Number:
Molecular Weight:
100.70
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

(single crystal substrate)

resistivity

~0.3 Ω-cm

diam. × thickness

2 in. × 0.5 mm

mp

1480 °C

density

4.13 g/mL at 25 °C

semiconductor properties

<111>

SMILES string

[P]#[Ga]

InChI

1S/Ga.P

InChI key

HZXMRANICFIONG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Physical properties

Thermal expansion: 5.3 x 10-6/°C
Undoped (N-type semiconductor), carrier concentration = 2-6 × 1016 cm-3, EPD < 3 × 105 cm-2, growth technique = LEC

Physical form

cubic (a = 5.4505Å)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santosh Khanal et al.
Nanomedicine : nanotechnology, biology, and medicine, 6(6), 707-713 (2010-07-06)
The tear film is a dynamic multilayered structure. The interactions and the interfacial dynamics between the layers that occur during a blink cycle must be such that they allow for maintenance of a stable tear film. Attempts to understand these
David Richards et al.
Langmuir : the ACS journal of surfaces and colloids, 26(11), 8141-8146 (2010-02-04)
Gallium phosphide is a semiconductor material that can be used for the fabrication of optoelectronic devices. The report compares the ability of two similar organic molecules to form covalent bonds with the GaP(100) surface. Undecenoic acid (UDA) is a terminal
Cecilia E Linsmeier et al.
Biomaterials, 29(35), 4598-4604 (2008-09-20)
Neural devices may play an important role in the diagnosis and therapy of several clinical conditions, such as stroke, trauma or neurodegenerative disorders, by facilitating motor and pain control. Such interfaces, chronically implanted in the CNS, need to be biocompatible
Marcos Paulo Pinheiro Ferreira et al.
Photomedicine and laser surgery, 27(6), 901-906 (2009-08-25)
To evaluate the effect of phototherapy on the viability of cultured C2C12 myoblasts under different nutritional conditions (muscle injury model) using low-energy gallium-aluminum-arsenide (GaAlAs) and aluminium-gallium-indium-phosphide (InGaAlP) lasers with different wavelengths and powers. The beneficial effects of phototherapy using low-energy
Ivan Avrutsky et al.
Optics express, 18(19), 20370-20383 (2010-10-14)
Integrated chip-scale optical systems are an attractive platform for the implementation of non-linear optical interactions as they promise compact robust devices that operate reliably with lower power consumption compared to analogs based on bulk nonlinear crystals. The use of guided

Articles

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Protocols

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Photoresist kit offers pre-weighed chemical components for lithographic processes, with separate etchants for various substrate choices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service