647756
1-Acetylpiperidine-4-carboxylic acid
97%
Synonym(s):
N-Acetylisonipecotic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H13NO3
CAS Number:
Molecular Weight:
171.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
180-184 °C (lit.)
functional group
carboxylic acid
SMILES string
CC(=O)N1CCC(CC1)C(O)=O
InChI
1S/C8H13NO3/c1-6(10)9-4-2-7(3-5-9)8(11)12/h7H,2-5H2,1H3,(H,11,12)
InChI key
WFCLWJHOKCQYOQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Reactant for synthesis of:
- Triazole derivatives
- Antiproliferative agents
- CDK inhibitors
- CCR5 antagonists for HIV treatment
- nhNK1 antagonists
- Piperidine derivatives
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service