Skip to Content
Merck
All Photos(2)

Key Documents

643009

Sigma-Aldrich

Cerium(IV) oxide, dispersion

greener alternative

nanoparticles, <25 nm particle size, 10 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CeO2
Molecular Weight:
172.11
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

nanoparticles

reaction suitability

reagent type: catalyst
core: cerium

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

10 wt. % in H2O

particle size

<25 nm

greener alternative category

SMILES string

O=[Ce]=O

InChI

1S/Ce.2O

InChI key

CETPSERCERDGAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is intended for Molecular Solar Thermal Energy Storage Systems (MOST) and has been enhanced for energy efficiency. Click here for more information.

Features and Benefits

No dispersant added

Legal Information

Product of Engi-Mat Co.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Anti-inflammatory Properties of Cerium Oxide Nanoparticles.
Hirst S, et al.
Small, 5(24), 2848-2856 (2009)
Pharmacological potential of cerium oxide nanoparticles
Celardo I, et al.
Nanoscale, 3(4), 1411-1420 null
Hailong Li et al.
Journal of hazardous materials, 243, 117-123 (2012-11-08)
MnO(x)-CeO(2) mixed-oxide supported on TiO(2) (Mn-Ce/Ti) was synthesized by an ultrasound-assisted impregnation method and employed to oxidize elemental mercury (Hg(0)) at 200°C in simulated coal combustion flue gas. Over 90% of Hg(0) oxidation was achieved on the Mn-Ce/Ti catalyst at
Ashraf Aly Hassan et al.
Journal of hazardous materials, 244-245, 251-258 (2012-12-29)
Experimental and computational investigation of the transport parameters of nanoparticles (NPs) flowing through porous media has been made. This work intends to develop a simulation applicable to the transport and retention of NPs in saturated porous media for investigating the
Cecilia Mondelli et al.
Chimia, 66(9), 694-698 (2012-12-06)
The heterogeneously catalyzed gas-phase oxidation of HCl to Cl(2) offers an energy-efficient and eco- friendly route to recover chlorine from HCl-containing byproduct streams in the chemical industry. This process has attracted renewed interest in the last decade due to an

Articles

As with all types of fuel cells, a Solid Oxide Fuel Cell (SOFC) is capable of efficiently transforming chemical energy into electrical energy.

Hydrogen is one of the most important resources in providing food, fuel, and chemical products for our everyday life. Sustainable catalytic hydrogen production from bioethanol has gained significant attention in recent years due to globally diminishing fossil fuel supplies, which have necessitated the search for new chemical feedstocks.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

See All

Questions

1–2 of 2 Questions  
  1. " <25 nm particle size"とありますが、測定方法を教えてください。分散液中のセリアを測定したか、乾燥状態のセリアを測定したかも含めてご開示いただけるとありがたいです。宜しくお願い致します。

    1 answer
    1. The dry particles are tested for BET surface area and that value is correlated back to an average particle size based on historical data and known relationships between these parameters. The particle size is not tested directly.

      Helpful?

  2. 通常cerium oxide nanoparticlesの水分散液は透明な黄緑色ですが、御社の製品であるcerium (IV) oxide, dispersion (643009)は不透明な白濁液です。可視光を遮断する為に白濁させている様ですが、何か保護剤は添加されておりますでしょうか?

    1 answer
    1. No protective agents are added to block visible light. The off-white appearance is simply due to the dispersion of the Cerium(IV) Oxide nanopowder in the water.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service