Skip to Content
Merck
All Photos(2)

Key Documents

64133

Sigma-Aldrich

1-Butyl-3-methylimidazolium bromide

dry, ≥98.5% (HPLC)

Synonym(s):

BMIMBr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15BrN2
CAS Number:
Molecular Weight:
219.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.5% (HPLC)

form

solid

impurities

≤500 ppm water

SMILES string

[Br-].CCCCn1cc[n+](C)c1

InChI

1S/C8H15N2.BrH/c1-3-4-5-10-7-6-9(2)8-10;/h6-8H,3-5H2,1-2H3;1H/q+1;/p-1

InChI key

KYCQOKLOSUBEJK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1-Butyl-3-methylimidazolium bromide is a neutral ionic liquid.

Application

BMIMBr undergoes gelation on reaction with gelatin to form ion jelly, a quasi-solid material for use in chemoresistive gas sensors. It can be used as a solvent for the preparation of 1,2,4,5-substituted imidazoles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Apparent molar volume and isentropic compressibility of ionic liquid 1-butyl-3-methylimidazolium bromide in water, methanol, and ethanol at T=(298.15 to 318.15) K
Zafarani-Moattar, Mohammed Taghi, and Hemayat Shekaari
The Journal of Chemical Thermodynamics, 37.10, 1029-1035 (2005)
Catalyst-free one-pot four component synthesis of polysubstituted imidazoles in neutral ionic liquid 1-butyl-3-methylimidazolium bromide.
Hasaninejad A, et al.
Journal of Combinatorial Chemistry, 12(6), 844-849 (2010)
Physical and electrochemical properties of 1-butyl-3-methylimidazolium bromide, 1-butyl-3-methylimidazolium iodide, and 1-butyl-3-methylimidazolium tetrafluoroborate
Kim, Ki-Sub, Bae-Kun Shin, and Huen Lee
Korean Journal of Chemical Engineering, 21.5, 1010-1014 (2004)
Density, viscosity and conductivity study of 1-butyl-3-methylimidazolium bromide
Tshibangu, Patrick Ngoy, Silindile Nomathemba Ndwandwe, and Ezekiel Dixon Dikio
International Journal of Electrochemistry, 6.6, 2-2 (2011)
Ion Jelly: a novel sensing material for gas sensors and electronic noses.
Carvalho T, et al.
Journal of Material Chemistry C, 2(4), 696-700 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service