Skip to Content
Merck
All Photos(3)

Key Documents

633801

Sigma-Aldrich

Calcium titanate

nanopowder, <100 nm particle size (BET), 99% trace metals basis

Synonym(s):

CALCIUM TITANIUM OXIDE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaTiO3
CAS Number:
Molecular Weight:
135.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99% trace metals basis

form

nanopowder

surface area

5-20 m2/g

particle size

<100 nm (BET)

density

4.1 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[O-][Ti]([O-])=O

InChI

1S/Ca.3O.Ti/q+2;;2*-1;

InChI key

AOWKSNWVBZGMTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matija Milanič et al.
Lasers in surgery and medicine, 45(1), 8-14 (2012-12-12)
Nd:YAP laser emitting at 1,342 nm appears promising for nonablative skin rejuvenation treatment, based on favorable absorption properties of water and melanin in this part of the spectrum. A quantitative determination of energy deposition characteristics of Nd:YAP in normal human skin
Annette Bussmann-Holder
Journal of physics. Condensed matter : an Institute of Physics journal, 24(27), 273202-273202 (2012-06-22)
This article reviews the polarizability model and its applications to ferroelectric perovskite oxides. The motivation for the introduction of the model is discussed and nonlinear oxygen ion polarizability effects and their lattice dynamical implementation outlined. While a large part of
Aiping Chen et al.
Advanced materials (Deerfield Beach, Fla.), 25(7), 1028-1032 (2012-11-28)
Intergrowth of two partially miscible phases of BiFeO(3) and BiMnO(3) gives a new class of room-temperature multiferroic phase, Bi(3) Fe(2) Mn(2) O(10+δ) , which has a unique supercell (SC) structure. The SC heterostructures exhibit simultaneously room-temperature ferrimagnetism and remanent polarization.
Y Wei et al.
Optics express, 20(9), 10399-10405 (2012-04-27)
We synthetize some new perovskite thin layers: p-fluorophenethylamine tetraiodoplumbate pFC(6)H(4)C(2)H(4)NH(3))(2)PbI(4) perovskite molecules, included in a PMMA matrix. We report on the optical properties of the perovskite doped PMMA thin layers and we show that these layers are much more stable
Wei Yang et al.
Environmental science & technology, 46(20), 11280-11288 (2012-09-19)
A series of LaFe(1-x)(Cu, Pd)(x)O(3-δ) perovskites was fully characterized and tested for the selective catalytic reduction (SCR) of NO by C(3)H(6) in the presence of O(2). The adsorbed species and surface reactions were investigated for mechanistic study by means of

Articles

Synthesis, Properties, and Applications of Perovskite-Phase Metal Oxide Nanostructures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service