Skip to Content
Merck
All Photos(1)

Key Documents

616702

Sigma-Aldrich

Sodium L-lactate-3,3,3-d3 solution

45-55 % (w/w) in H2O, ≥98 atom % D, ≥98% (CP), ≥98% (Chiral Purity, HPLC)

Synonym(s):

L-Lactic-3,3,3-d3 acid sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CH(OH)CO2Na
CAS Number:
Molecular Weight:
115.08
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

description

Packaged as gram of solution, 45-55% (w/w)

isotopic purity

≥98 atom % D

Assay

≥98% (CP)
≥98% (Chiral Purity, HPLC)

concentration

45-55 % (w/w) in H2O

technique(s)

mass spectrometry (MS): suitable

mp

163-165 °C (lit.)

mass shift

M+3

storage temp.

−20°C

SMILES string

[Na+].[2H]C([2H])([2H])[C@H](O)C([O-])=O

InChI

1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1/i1D3;

InChI key

NGSFWBMYFKHRBD-QQAVFRBQSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service