608149
L-Methionine-(carboxy-13C,methyl-d3)
99 atom % D, 99 atom % 13C
Synonym(s):
SILAC Amino Acid
About This Item
Recommended Products
isotopic purity
99 atom % 13C
99 atom % D
form
solid
optical activity
[α]25/D +23.1°, c = 1 in 1 M HCl
technique(s)
protein expression: suitable
mp
284 °C (dec.) (lit.)
mass shift
M+4
SMILES string
[2H]C([2H])([2H])SCC[C@H](N)[13C](O)=O
InChI
1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1/i1D3,5+1
InChI key
FFEARJCKVFRZRR-YLXZAIPNSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service