Skip to Content
Merck
All Photos(1)

Key Documents

606502

Sigma-Aldrich

Glyphosate-2-13C

99 atom % 13C

Synonym(s):

N-(Phosphonomethyl)glycine-2-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C13CH2NHCH2PO(OH)2
CAS Number:
Molecular Weight:
170.07
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

mp

230 °C (dec.) (lit.)

application(s)

agriculture
environmental

mass shift

M+1

SMILES string

OC(=O)[13CH2]NCP(O)(O)=O

InChI

1S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)/i1+1

InChI key

XDDAORKBJWWYJS-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valerio Matozzo et al.
Fish & shellfish immunology, 83, 321-329 (2018-09-16)
The effects of the breakdown products of herbicides on aquatic species are largely unknown. In a recent study, we evaluated the effects of glyphosate on the mussel Mytilus galloprovincialis. This study was designed to evaluate for the first time the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service