All Photos(1)
About This Item
Linear Formula:
84Kr
CAS Number:
Molecular Weight:
83.91
MDL number:
UNSPSC Code:
12142003
PubChem Substance ID:
Recommended Products
isotopic purity
80 atom %
Assay
≥99.5% (Total Krypton)
form
gas
impurities
<0.5% Total others
mass shift
depleted
SMILES string
[84Kr]
InChI
1S/Kr/i1+0
InChI key
DNNSSWSSYDEUBZ-IGMARMGPSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Packaging
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Press. Gas Compr. Gas
Storage Class Code
2A - Gases
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Xavier Devaux et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 16(12), 2692-2701 (2015-07-03)
The results of a study on the evolution of HiPco single-walled carbon nanotubes during the oxidizing action of H2 SO4 and HNO3 are presented. The process conditions used have been chosen so as to avoid any significant damage to the
Gamil A Guirgis et al.
The journal of physical chemistry. A, 119(47), 11532-11547 (2015-10-06)
The FT-microwave spectrum (6.5-26 GHz) of (chloromethyl)fluorosilane (ClCH2-SiH2F) has been recorded and 250 transitions for the parent species along with (13)C, (37)Cl, (29)Si, and (30)Si isotopologues have been assigned for trans conformer. Infrared spectra (3100 to 400 cm(-1)) of gas
James G Carter et al.
Angiogenesis, 18(1), 23-30 (2014-10-03)
Anti-VEGF-A therapy has become a mainstay of treatment for ocular neovascularisation and in cancer; however, their effectiveness is not universal, in some cases only benefiting a minority of patients. Anti-VEGF-A therapies bind and block both pro-angiogenic VEGF-Axxx and the partial
Seth H Pincus et al.
Toxins, 7(6), 2121-2133 (2015-06-13)
Ricin is a potential bioweapon that could be used against civilian and military personnel. Aerosol exposure is the most likely route of contact to ricin toxin that will result in the most severe toxicity. Early recognition of ricin exposure is
Yvonne Kohlmann et al.
Journal of proteome research, 13(10), 4325-4338 (2014-09-10)
Ralstonia eutropha H16 is a denitrifying microorganism able to use nitrate and nitrite as terminal electron acceptors under oxygen deprivation. To identify proteins showing an altered expression pattern in response to oxygen supply, R. eutropha cells grown aerobically and anaerobically
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service