Skip to Content
Merck
All Photos(3)

Key Documents

594849

Sigma-Aldrich

Titanium(IV) carbide

powder, <4 μm, ≥99% (Ti)

Synonym(s):

TiC-HP, Titanium monocarbide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiC
CAS Number:
Molecular Weight:
59.88
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99% (Ti)

form

powder

particle size

<4 μm

bp

4820 °C (lit.)

mp

3140 °C (lit.)

density

4.930 g/mL at 25 °C (lit.)

SMILES string

[C-]#[Ti+]

InChI

1S/C.Ti/q-1;+1

InChI key

YXIVWSJCLXKLJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Rohrer et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 22(1), 015004-015004 (2010-01-13)
We present a strategy to identify energetically favourable oxide structures in thin-film geometries. Thin-film candidate configurations are constructed from a pool of sublattices of stable and metastable oxide bulk phases. Favourable stoichiometric compositions and atomic geometries are identified by comparing
Marina Brama et al.
Biomaterials, 28(4), 595-608 (2006-10-20)
Titanium has limitations in its clinical performance in dental and orthopaedic applications. This study describes a coating process using pulsed laser deposition (PLD) technology to produce surfaces of titanium carbide (TiC) on titanium substrates and evaluates the biological response both
Hiroshi Moriwaki et al.
Journal of hazardous materials, 185(2-3), 725-731 (2010-10-26)
The aim of this study is to investigate the utilization of the powder of porous titanium carbide (TiC) ceramics as a novel adsorbent or a material for solid-phase extraction (SPE). The adsorption and elution of inorganic and organic pollutants, Pb(II)
Raimo Silvennoinen et al.
Optics express, 16(14), 10130-10140 (2008-07-09)
Adsorption of human plasma fibrinogen (HPF) on 6 differently treated titanium samples (polished, polished and etched, and 4 titanium carbide coatings samples produced by using plasma-enhanced chemical vapour deposition (PECVD) method) is investigated by using diffractive optical element (DOE) sensor.
Yuhe Zhu et al.
Dental materials journal, 26(2), 245-253 (2007-07-12)
Carbide layer was formed on the surface of Ti by heating in hydrocarbon atmosphere (benzene C6H6) at 1000-1400 degrees C using a high frequency induction heating method. Physical and mechanical properties of carbide-coated Ti were investigated to examine its potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service