All Photos(1)
About This Item
Linear Formula:
HC≡CC6H4CHO
CAS Number:
Molecular Weight:
130.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
64-67 °C (lit.)
functional group
aldehyde
SMILES string
[H]C(=O)c1ccccc1C#C
InChI
1S/C9H6O/c1-2-8-5-3-4-6-9(8)7-10/h1,3-7H
InChI key
ZEDSAJWVTKUHHK-UHFFFAOYSA-N
Application
2-Ethynylbenzaldehyde may be used in the synthesis of the following:
- iodoisoquinoline-fused benzimidazoles obtained via tandem iodocyclization of 2-ethynylbenzaldehyde with o-benzenediamine and iodine in the presence of copper(I)iodide
- N-[(7,7a-dihydroisoquinolino[2,1-a]perimidin-13-yl)methyl]-N-isopropylpropan-2-amine obtained via a multi-step process
- 2-[(2-bromophenyl)ethynyl]benzaldehyde
- 2-[(2-bromo-5-fluorophenyl)ethynyl]benzaldehyde
- 2-[(2-bromo-5-methylphenyl)ethynyl]benzaldehyde
- 2-(phenylethynyl)benzaldehyde
- 2-[(4-methylphenyl)ethynyl]benzaldehyde
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
?CuI/I2-Promoted Electrophilic Tandem Cyclization of 2 Ethynylbenzaldehydes with ortho-Benzenediamines: Synthesis of
Iodoisoquinoline-Fused Benzimidazoles?
Iodoisoquinoline-Fused Benzimidazoles?
Ouyang C-H, et al.
The Journal of Organic Chemistry, 76(01), 223-228 (2010)
?Direct synthesis of highly fused perimidines by copper(I)-catalyzed
hydroamination of 2-ethynylbenzaldehydes?
hydroamination of 2-ethynylbenzaldehydes?
Tokimizu Y, et al.
Tetrahedron, 67(29), 5168- 5175 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service