Skip to Content
Merck
All Photos(1)

Key Documents

586846

Sigma-Aldrich

N-Benzoyladenosine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N5O5
CAS Number:
Molecular Weight:
371.35
Beilstein:
632501
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

optical activity

[α]20/D -49°, c = 1% in DMSO

mp

141-145 °C (lit.)

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(NC(=O)c4ccccc4)ncnc23

InChI

1S/C17H17N5O5/c23-6-10-12(24)13(25)17(27-10)22-8-20-11-14(18-7-19-15(11)22)21-16(26)9-4-2-1-3-5-9/h1-5,7-8,10,12-13,17,23-25H,6H2,(H,18,19,21,26)/t10-,12-,13-,17-/m1/s1

InChI key

NZDWTKFDAUOODA-CNEMSGBDSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service