579009
Poly(Disperse Red 1 methacrylate)
Synonym(s):
Poly[4′-[[2-(methacryloxy)ethyl]ethyl]amino-4-nitroazobenzene]
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
mp
>300 °C (lit.)
Quality Level
transition temp
Tg 82 °C (onset)
solubility
chloroform: soluble
toluene: soluble
λmax
467 nm
SMILES string
[N+](=O)([O-])c1ccc(cc1)N=Nc2ccc(cc2)N(CCOC(=O)C(=C)C)CC
InChI key
MHYWMAUCJNDHPY-UHFFFAOYSA-N
Related Categories
General description
NLO polymer
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Scientific reports, 10(1), 3267-3267 (2020-02-26)
The isomerisation of azo dyes can induce conformational changes which have potential applications in medicine and environmental protection. We developed an agar diffusion assay to test the capture and release of biologically active molecules from an azo electro-optic polymer, Poly
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service