Skip to Content
Merck
All Photos(1)

Key Documents

56655

Sigma-Aldrich

(−)-Methyl (R)-3-hydroxyvalerate

≥98.0% (sum of enantiomers, GC)

Synonym(s):

(−)-Methyl (R)-3-hydroxypentanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O3
CAS Number:
Molecular Weight:
132.16
Beilstein:
4655380
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (sum of enantiomers, GC)

optical activity

[α]20/D −37±3°, c = 1% in chloroform

refractive index

n20/D 1.427

bp

68-70 °C/5 mmHg (lit.)

density

1.029 g/mL at 20 °C (lit.)

functional group

ester
hydroxyl

SMILES string

CC[C@@H](O)CC(=O)OC

InChI

1S/C6H12O3/c1-3-5(7)4-6(8)9-2/h5,7H,3-4H2,1-2H3/t5-/m1/s1

InChI key

XHFXKKFVUDJSPJ-RXMQYKEDSA-N

Application

  • Preparation of single-enantiomer 2-methyl-4-heptanol, a pheromone of Metamasius hemipterus, using (S)-2-methoxy-2-(1-naphthyl)propionic acid.: This research outlines the synthesis of a single-enantiomer pheromone compound using (S)-2-methoxy-2-(1-naphthyl)propionic acid. The process highlights the importance of enantiopure intermediates like (−)-Methyl (R)-3-hydroxyvalerate in the synthesis of biologically active compounds (Ichikawa and Ono, 2006).

Other Notes

Chiral building block for EPC syntheses; selective anti-alkylation of its dianion; OH protection and reduction of ester group to aldehyde or alcohol functions; stereoselective preparation of a dioxanone with pivalaldehyde

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Mori et al.
Liebigs Ann. Chem., 159-159 (1990)
K. Mori et al.
Tetrahedron, 41, 919-919 (1985)
G. Frater
Helvetica Chimica Acta, 62, 2825-2825 (1979)
P. DeShong et al.
Tetrahedron Letters, 27, 2091-2091 (1986)
Diya Alsafadi et al.
New biotechnology, 34, 47-53 (2016-05-26)
Olive mill wastewater (OMW), a highly polluting waste from the olive oil industry, was utilized as sole carbon source for the production of polyhydroxyalkanoate (PHA) by extremely halophilic Haloferax Mediterranei (H. mediterranei) in a one stage cultivation step. H. mediterranei

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service