All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H6N2S
CAS Number:
Molecular Weight:
150.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
mp
87-91 °C (lit.)
SMILES string
Nc1ccc2ncsc2c1
InChI
1S/C7H6N2S/c8-5-1-2-6-7(3-5)10-4-9-6/h1-4H,8H2
InChI key
FAYAYUOZWYJNBD-UHFFFAOYSA-N
General description
6-Aminobenzothiazole can be prepared from 6-nitrobenzothiazole via sonochemical reduction method.
Application
6-Aminobenzothiazole may be used in the synthesis of following:
- 1-(6-amino-benzothiazolyl)-3-chloro-5-methoxytriazine
- 6-[(4-N,N-dimethylaminophenyl)diazenyl]benzothiazole
- 6-[(2-hydroxy-1-naphthyl)diazenyl]benzothiazole
- 6-dimethylaminobenzothiazole
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
?The synthesis and structural study of two benzothiazolyl azo dyes: X-ray crystallographic and computational study of azo?hydrazone tautomerism?
Pavlovic G, et al.
Dyes and Pigments, 83(3), 354-362 (2009)
"Preparation of novel push-pull benzothiazole derivatives with reverse polarity: compounds with potential non-linear optic application"
Hrobarik P, et al.
Synthesis, 2005(04), 600-604 (2005)
"2-Hydroxy-4, 6-diamino-[1, 3, 5] triazines: a novel class of VEGF-R2 (KDR) tyrosine kinase inhibitors"
Baindur N, et al.
Journal of Medicinal Chemistry, 48(6), 1717-1720 (2005)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service