Skip to Content
Merck
All Photos(1)

Key Documents

55332

Sigma-Aldrich

Luviquat FC 550

~40% active ingredients in H2O

Synonym(s):

Poly[(3-methyl-1-vinylimidazolium chloride)-co-(1-vinylpyrrolidone)], Polyquaternium D16

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

description

cationic

concentration

~40% active ingredients in H2O

technique(s)

protein purification: suitable
protein quantification: suitable

refractive index

n20/D 1.417

pH

5.0-8.0 (10% in H2O, active ingredients)

density

1.11 g/mL at 20 °C

SMILES string

[Cl-].[n+]2(c[n](cc2)C)C=C.N1(CCCC1=O)C=C

InChI

1S/C6H9N2.C6H9NO.ClH/c1-3-8-5-4-7(2)6-8;1-2-7-5-3-4-6(7)8;/h3-6H,1H2,2H3;2H,1,3-5H2;1H/q+1;;/p-1

InChI key

ZIQRJGXRRBOCEI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Luviquat is a trademark of BASF SE

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service