Skip to Content
Merck
All Photos(1)

Documents

544922

Sigma-Aldrich

Ammonium hexachloroosmate(IV)

99.99% trace metals basis

Synonym(s):

Ammonium osmium chloride, Osmium(IV)-ammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2OsCl6
CAS Number:
Molecular Weight:
439.02
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

powder

impurities

≤150.0 ppm Trace Metal Analysis

mp

170 °C (subl.) (lit.)

density

2.93 g/mL at 25 °C (lit.)

SMILES string

[NH4+].[NH4+].Cl[Os--](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.2H3N.Os/h6*1H;2*1H3;/q;;;;;;;;+4/p-4

InChI key

SRBXXQDKBKTWOC-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chee Shan Lim et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 16(9), 1898-1905 (2015-04-25)
Renewable energy sources are highly sought after as a result of numerous worldwide problems concerning the environment and the shortage of energy. Currently, the focus in the field is on the development of catalysts that are able to provide water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service