Skip to Content
Merck
All Photos(1)

Documents

544000

Sigma-Aldrich

5-Hexynoic acid

97%

Synonym(s):

4-Ethynylbutyric acid, 5-Hexyn-1-oic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH≡C(CH2)3CO2H
CAS Number:
Molecular Weight:
112.13
Beilstein:
1743192
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.449 (lit.)

bp

224-225 °C (lit.)

density

1.016 g/mL at 25 °C (lit.)

SMILES string

OC(=O)CCCC#C

InChI

1S/C6H8O2/c1-2-3-4-5-6(7)8/h1H,3-5H2,(H,7,8)

InChI key

VPFMEXRVUOPYRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

5-Hexynoic acid is an alkynoic acid. It undergoes gold-catalyzed cyclization to form the corresponding methylene lactones.

Application

5-Hexynoic acid may be used in the preparation of:
  • 5-hexynoic acid methyl ester
  • 5,6-dicarba-closo-dodecaboranyl hexynoic acid
  • 5,8,11-dodecatriynoic acid
  • 5-isocyanato-1-pentyne

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A mild access to ?-or ?-alkylidene lactones through gold catalysis.
Harkat H, et al.
Tetrahedron Letters, 47(35), 6273-6276 (2006)
Juan M Castro et al.
Scientific reports, 9(1), 165-165 (2019-01-19)
The self-reproduction of supramolecular assemblies based on the synthesis and self-assembly of building blocks is a critical step towards the construction of chemical systems with autonomous, adaptive, and propagation properties. In this report, we demonstrate that giant vesicles can grow
Cobalt-catalyzed cocyclizations of isocyanato alkynes: a regiocontrolled entry into 5-indolizinones. Application to the total synthesis of camptothecin.
Earl RA and Vollhardt KPC.
Journal of the American Chemical Society, 105(23), 6991-6993 (1983)
Synthesis of 5, 8, 11-dodecatriynoic acid and its use in the synthesis of arachidonic acid and related acids.
Fryer RI, et al.
The Journal of Organic Chemistry, 40(3), 348-350 (1975)
Synthesis of a Tyr3-octreotate conjugated closo-carborane [HC2B10H10]: a potential compound for boron neutron capture therapy.
Schirrmacher E, et al.
Tetrahedron Letters, 44(51), 9143-9145 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service