541567
Methyl 5-(chloromethyl)-2-furoate
97%
Synonym(s):
Methyl 5-(chloromethyl)-2-furate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C7H7ClO3
CAS Number:
Molecular Weight:
174.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
mp
32-36 °C (lit.)
storage temp.
2-8°C
SMILES string
COC(=O)c1ccc(CCl)o1
InChI
1S/C7H7ClO3/c1-10-7(9)6-3-2-5(4-8)11-6/h2-3H,4H2,1H3
InChI key
PWXMEBZOKUPCST-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis and Reactions of Substituted 3-amino-2-furyl (aryl)-thieno [2, 3-b] pyridines.
Kaigorodova YA, et al.
Molecules (Basel), 5(10), 1085-1093 (2000)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service