Skip to Content
Merck
All Photos(1)

Key Documents

540285

Sigma-Aldrich

Tetraalkylammonium carbonate, polymer-bound

macroporous, 18-50 mesh, extent of labeling: 2.5-3.5 mmol/g N loading

Synonym(s):

MP-carbonate resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

form

macroporous

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: proton reactive

extent of labeling

2.5-3.5 mmol/g N loading

particle size

18-50 mesh

SMILES string

[O-]C([O-])=O.C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.CC[N+](CC)(CC)Cc3ccccc3.CC[N+](CC)(CC)Cc4ccccc4

InChI key

RBIYTBCFDIQNIC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Tetraalkylammonium carbonate, polymer-bound also known as MP-carbonate resin, is a strong basic macroporous anion-exchange polystyrene resin that is commonly used in solution phase peptide synthesis and resin-based purification techniques.

Application

Tetraalkylammonium carbonate, polymer-bound, is used as a solid support base in the solution phase parallel synthesis of amides to remove unwanted acidic by-products. It is used as a solid-phase base to neutralize the hydrobromide in the synthesis of syringolin B.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses and cytotoxicity of syringolin B-based proteasome inhibitors
TR Ibarra-Rivera
Tetrahedron, 67, 9950-9956 (2011)
Solution-phase parallel synthesis of 5-carboxamido 1-benzyl-3-(3-dimethylaminopropyloxy)-1H-pyrazoles as activators of soluble guanylate cyclase with improved oral bioavailability
DL Selwood
Bioorganic & Medicinal Chemistry Letters, 11, 1089-1092 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service