Skip to Content
Merck
All Photos(1)

Key Documents

524433

Sigma-Aldrich

N-Benzylcinchonidinium bromide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29BrN2O
CAS Number:
Molecular Weight:
465.43
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

Quality Level

Assay

97%

optical activity

[α]20/D −138°, c = 1 in chloroform

mp

190 °C (dec.) (lit.)

InChI key

UUVFTQCFSNVLGV-SPTWEYDNSA-M

General description

N-Benzylcinchonidinium bromide is a chiral phase-transfer catalyst (PTC).

Application

N-Benzylcinchonidinium bromide (BCDBr) can be used:
  • To prepare various 4-(bromomethyl)benzenesulfonamides, which are employed as catalysts for asymmetric benzylation reactions.
  • As a phase transfer catalyst in the synthesis of organogelators via Michael addition reaction.
  • To catalyze the enantioselective alkylation of tert-butyl glycinate-benzophenone Schiff base furnishing the corresponding chiral α-amino acid.
  • To resolve symmetric biaryldiols via formation of inclusion complex with diols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective reactions of tert-butyl glycinate-benzophenone Schiff base catalyzed by chiral phase-transfer catalyst in aqueous media without any organic solvent.
Mase N, et al.
Tetrahedron Letters, 46(18), 3213-3216 (2005)
Synthesis of cinchonidinium salts containing sulfonamide functionalities and their catalytic activity in asymmetric alkylation reactions
Itsuno S, et al.
Tetrahedron Letters, 55(44), 6117-6120 (2014)
Synthesis of green organogelators with a sulfide linkage via solvent-free Michael addition: soft templates for the preparation of size-controlled gold nanoparticles
Delbecq F, et al.
Tetrahedron Letters, 54(7), 651-656 (2013)
Separation of the Enantiomers of 2, 2?-Dihydroxy-1, 1?-binaphthyl and 10, 10?-Dihydroxy-9, 9?-biphenanthryl by Complexation with N-Alkylcinchonidinium Halides
Tanaka K, et al.
Angewandte Chemie (International ed. in English), 32(8), 1147-1148 (1993)

Articles

Maruoka catalysts enable enantioselective preparation of α-amino acids from glycine derivatives via asymmetric phase transfer catalysis.

Maruoka catalysts enable enantioselective preparation of α-amino acids from glycine derivatives via asymmetric phase transfer catalysis.

Maruoka catalysts enable enantioselective preparation of α-amino acids from glycine derivatives via asymmetric phase transfer catalysis.

Maruoka catalysts enable enantioselective preparation of α-amino acids from glycine derivatives via asymmetric phase transfer catalysis.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service