Skip to Content
Merck
All Photos(1)

Key Documents

51330

Sigma-Aldrich

Harmaline

≥95%

Synonym(s):

1-Methyl-7-methoxy-3,4-dihydro-β-carboline, 3,4-Dihydroharmine, 4,9-Dihydro-7-methoxy-1-methyl-3H-pyrido[3,4-b]indole, Dihydroharmine, Harmalol methyl ether, Harmidine, NSC 407285

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14N2O
CAS Number:
Molecular Weight:
214.26
Beilstein:
207310
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada
stupéfiant (France); regulated under CDSA - not available from Sigma-Aldrich Canada

mp

232-234 °C (lit.)

SMILES string

COc1ccc2c3CCN=C(C)c3[nH]c2c1

InChI

1S/C13H14N2O/c1-8-13-11(5-6-14-8)10-4-3-9(16-2)7-12(10)15-13/h3-4,7,15H,5-6H2,1-2H3

InChI key

RERZNCLIYCABFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Acylated harmalines via Friedel-Crafts reaction
  • Phenacyl derivatives of 1-methyl-7-methoxy-β-carbolines as central nervous system affectors
  • Palladium harmaline DMSO chloro complex as anti-tumor agent

Natural product scaffold in preparation of:
  • Trypanothione reductase inhibitors†

Biochem/physiol Actions

CNS stimulant; may act through NMDA receptors

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 2

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hamid-Reza Adhami et al.
Phytotherapy research : PTR, 25(8), 1148-1152 (2011-02-03)
To find new herbal compounds with an acetylcholinesterase (AChE) inhibitory effect, this study focused on herbal drugs and resins which have been used in Iranian traditional medicine for the treatment of cognitive disorders. Forty drugs were selected from authoritative written
Muhammad Rizwan-ul-Haq et al.
Chemosphere, 85(6), 995-1001 (2011-07-26)
Apolipophorin III, traditionally known for lipid transport in insects is fairly established as toxicity indicator against harmaline and tea saponin during this study. Apolipophorin III expressed in the hemolymph and midgut tissues of 3rd, 4th, 5th larval instars and pupae
Pervin K Iseri et al.
Neuropharmacology, 61(4), 715-723 (2011-06-07)
Essential tremor (ET) is one of the most common and most disabling movement disorders among adults. The drug treatment of ET remains unsatisfactory. Additional therapies are required for patients with inadequate response or intolerable side effects. The current study aims
Fatta B Nahab et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 9(3), 635-638 (2012-03-29)
Recent work exploring the use of high-molecular weight alcohols to treat essential tremor (ET) has identified octanoic acid as a potential novel tremor-suppressing agent. We used an established harmaline-based mouse model of ET to compare tremor suppression by 1-octanol and
Jordi Riba et al.
Drug testing and analysis, 4(7-8), 610-616 (2012-04-20)
Ayahuasca is an Amazonian psychotropic plant tea obtained from Banisteriopsis caapi, which contains β-carboline alkaloids, chiefly harmine, harmaline and tetrahydroharmine. The tea usually incorporates the leaves of Psychotria viridis or Diplopterys cabrerana, which are rich in N,N-dimethyltryptamine (DMT), a psychedelic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service