Skip to Content
Merck
All Photos(1)

Key Documents

510653

Sigma-Aldrich

Copper(I) sulfide

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Chalcocite, Copper(1+) sulfide, Dicopper monosulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu2S
CAS Number:
Molecular Weight:
159.16
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Assay

99.99% trace metals basis

form

powder

technique(s)

NMR: suitable

impurities

≤150.0 ppm Trace Metal Analysis

density

5.6 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Cu]S[Cu]

InChI

1S/2Cu.S

InChI key

JESPAFKOCOFQIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cu63 NMR, X ray photoelectron spectroscopy,2 copper sulfide CuS was studied in detail. Copper sulfide is a monovalent and oxidation state was determined to be 2.2

Packaging

Packaged in ampules

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aaron E Saunders et al.
Nano letters, 6(12), 2959-2963 (2006-12-14)
The self-assembly of sterically stabilized colloidal copper sulfide nanodisks, 14-20 nm in diameter and 5-7 nm thick, was studied. The nanodisks were observed by electron microscopy and small-angle X-ray scattering to form columnar arrays when evaporated as thin films from
Kanishka Biswas et al.
Journal of colloid and interface science, 333(1), 404-410 (2009-02-24)
The interface between toluene and water has been employed to prepare ultrathin Janus nanocrystalline films of metal oxides, metal chalcogenides and gold, wherein the surface on the organic-side is hydrophobic and the aqueous-side is hydrophilic. We have changed the nature
Shusheng Zhang et al.
Analytical chemistry, 80(19), 7206-7212 (2008-09-02)
A novel and sensitive flow injection chemiluminescence assay for sequence-specific DNA detection based on signal amplification with nanoparticles (NPs) is reported in the present work. The "sandwich-type" DNA biosensor was fabricated with the thiol-functionalized capture DNA first immobilized on an
Vinod Kumar Gupta et al.
Journal of hazardous materials, 243, 179-186 (2012-11-06)
This study describes the effect of adsorption on methylene blue degradation using pectin-CuS nanocomposite (PCSNC). The nanocomposite was synthesized using co-precipitation methods followed by direct encapsulation with pectin. The synthesized nanocomposite was characterized by SEM, TEM, XRD, FTIR and UV-vis
T P Mokone et al.
Journal of colloid and interface science, 351(1), 10-18 (2010-08-14)
Metal sulfide precipitation forms an important component of acid mine drainage remediation systems based on bacterial sulfate reduction. The precipitation reaction is thermodynamically favorable, but a number of technical issues remain. In this study the effect of metal to sulfide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service