50893
1-Butyl-1-methylpyrrolidinium dicyanamide
≥97.0% (T)
Synonym(s):
[BMPYR][DCA]
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H20N4
CAS Number:
Molecular Weight:
208.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥97.0% (T)
impurities
≤0.5% water
functional group
amine
SMILES string
N#C[N-]C#N.CCCC[N+]1(C)CCCC1
InChI
1S/C9H20N.C2N3/c1-3-4-7-10(2)8-5-6-9-10;3-1-5-2-4/h3-9H2,1-2H3;/q+1;-1
InChI key
LBHLGZNUPKUZJC-UHFFFAOYSA-N
General description
1-Butyl-1-methylpyrrolidinium dicyanamide is a hypergolic, air- and water-stable pyrrolidinium-based ionic liquid. It shows a wide potential range, making it a promising candidate in electrochemical applications. [BMPYR][DCA] also has high extraction capacity for dibenzothiophene from liquid fuels.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Voltammetric Study and Electrodeposition of Ni (II)/Fe (II) in the Ionic Liquid 1-Butyl-1-Methylpyrrolidinium Dicyanamide
Lo, Nai-Chang, et al.
Journal of the Electrochemical Society, 163.2, D9-D16 (2016)
Ionic liquids as hypergolic fuels.
Schneider S, et al.
Energy and Fuels, 22(4), 2871-2871 (2008)
Thermophysical properties of the pure ionic liquid 1-butyl-1-methylpyrrolidinium dicyanamide and its binary mixtures with alcohols
Gonza?lez, Emilio J., Begon?a Gonza?lez, and Euge?nia A. Macedo
Journal of Chemical and Engineering Data, 58.6, 1440-1448 (2013)
Electrodeposition of gold thin films from 1-butyl-1-methylpyrrolidinium dicyanamide Au 3+ solutions
De Sa, A. I., et al.
Thin Solid Films, 519.19, 6278-6283 (2011)
Pyrrolidinium-based ionic liquids. 1-Butyl-1-methyl pyrrolidinium dicyanoamide: thermochemical measurement, mass spectrometry, and ab initio calculations.
Emel'yanenko VN, et al.
The Journal of Physical Chemistry B, 112(37), 11734-11742 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service