49637
1-Methyl-3-propylimidazolium iodide
≥98.0% (HPLC)
Synonym(s):
MPII
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥98.0% (HPLC)
impurities
≤0.3% water
≤50 mg/kg chloride and bromide
SMILES string
[I-].CCC[n+]1ccn(C)c1
InChI
1S/C7H13N2.HI/c1-3-4-9-6-5-8(2)7-9;/h5-7H,3-4H2,1-2H3;1H/q+1;/p-1
InChI key
IVCMUVGRRDWTDK-UHFFFAOYSA-M
General description
1-Methyl-3-propylimidazolium iodide can be prepared by reacting 1-methylimidazole and propyl iodide.
Application
MPII in combination with poly(vinylidenefluoride-co-hexafluoropropylene) (PVDF-HFP) forms an ionic liquid polymer gel electolyte, which can be used in dye-sensitized nanocrystalline titanium dioxide (TiO2) solar cells.
Non-aqueous electrolyte for dye-sensitized solar cells.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nanoscale research letters, 6(1), 307-307 (2011-06-30)
The presence of specific chemical additives in the redox electrolyte results in an efficient increase of the photovoltaic performance of dye-sensitized solar cells (DSCs). The most effective additives are 4-tert-butylpyridine (TBP), N-methylbenzimidazole (NMBI) and guanidinium thiocyanate (GuNCS) that are adsorbed
Langmuir : the ACS journal of surfaces and colloids, 32(6), 1521-1532 (2016-01-23)
A library of six selenorhodamine dyes (4-Se-9-Se) were synthesized, characterized, and evaluated as photosensitizers of TiO2 in dye-sensitized solar cells (DSSCs). The dyes were constructed around either a bis(julolidyl)- or bis(half-julolidyl)-modified selenoxanthylium core functionalized at the 9-position with a thienyl
Influence of various cations on redox behavior of I- and I 3- and comparison between KI complex with 18-crown-6 and 1, 2-dimethyl-3-propylimidazolium iodide in dye-sensitized solar cells.
Electrochimica Acta, 50(13), 2597-2602 (2005)
High efficiency dye-sensitized nanocrystalline solar cells based on ionic liquid polymer gel electrolyte.
Chemical Communications (Cambridge, England), 24, 2972-2973 (2002)
Chemical communications (Cambridge, England), (24), 2972-2973 (2003-01-23)
An ionic liquid polymer gel containing 1-methyl-3-propylimidazolium iodide (MPII) and poly(vinylidenefluoride-co-hexafluoropropylene) (PVDF-HFP) has been employed as quasi-solid-state electrolyte in dye-sensitized nanocrystalline TiO2 solar cells with an overall conversion efficiency of 5.3% at AM 1.5 illumination.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service