Skip to Content
Merck
All Photos(1)

Documents

494534

Sigma-Aldrich

Cobalt(III) acetylacetonate

99.99% trace metals basis

Synonym(s):

Co(acac)3, Cobalt(III) 2,4-pentanedionate, Cobaltic acetylacetonate, Tris(2,4-pentanedionato)cobalt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Co(C5H7O2)3
CAS Number:
Molecular Weight:
356.26
Beilstein:
9007210
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

granular
powder or crystals

reaction suitability

core: cobalt

mp

210-213 °C (lit.)

SMILES string

CC(=O)\C=C(\C)O[Co](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.Co/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-;

InChI key

RHCQEPWEBDOALW-LNTINUHCSA-K

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Used for the preparation of a new family of monoporphyrinates which are interesting for their photochemical and magnetic properties and synthetic applications.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gema Martinez et al.
Frontiers in chemistry, 6, 487-487 (2018-11-06)
C-encapsulated highly pure PtxCoy alloy nanoparticles have been synthesized by an innovative one-step in-situ laser pyrolysis. The obtained X-ray diffraction pattern and transmission electron microscopy images correspond to PtxCoy alloy nanoparticles with average diameters of 2.4 nm and well-established crystalline
I K Adzamli et al.
The Biochemical journal, 211(1), 219-226 (1983-04-01)
Kinetic results are presented for the reaction of reduced [2Fe-2S] ferredoxin from the blue-green alga Spirulina platensis with Co(NH3)6(3+), Co(edta)- and Co(acac)3 as oxidants at pH 8.0 at I0.10 (NaCl). The aim is to compare results obtained with those previously
Akira Ohashi et al.
Talanta, 146, 789-794 (2015-12-24)
The solubilities of several cobalt(III) chelates (CoL3) with in supercritical carbon dioxide (SC-CO2) were investigated in the presence of fluorine- and trifluoromethyl-substituted phenols (PhOH) using UV-vis spectrophotometry. Solubility enhancement of CoL3 complexes in SC-CO2 was accomplished by adding PhOH. The

Articles

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service