Skip to Content
Merck
All Photos(1)

Key Documents

494135

Sigma-Aldrich

Tin(IV) tert-butoxide

≥99.99% trace metals basis

Synonym(s):

Tin tetra(tert -butoxide), Tin(IV) tert -butoxide, Tin(IV) tetra-tert -butoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sn(OC(CH3)3)4
CAS Number:
Molecular Weight:
411.16
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99% trace metals basis

form

low-melting solid

reaction suitability

core: tin

mp

40-44 °C (lit.)

SMILES string

CC(C)(C)O[Sn](OC(C)(C)C)(OC(C)(C)C)OC(C)(C)C

InChI

1S/4C4H9O.Sn/c4*1-4(2,3)5;/h4*1-3H3;/q4*-1;+4

InChI key

OSXGKVOYAKRLCS-UHFFFAOYSA-N

Application

  • Metal Oxide Electrospun Nanofibrous Membranes for Effective Dye Degradation and Sustainable Photocatalysis: This study involves the use of Tin(IV) tert-butoxide in the preparation of metal oxide nanofibrous membranes for photocatalytic applications, highlighting its relevance in material science and sustainable technology (V Jagadeesh Babu et al., 2023).
  • Atmospheric atomic layer deposition of SnO2 thin films with tin (II) acetylacetonate and water: The research uses Tin(IV) tert-butoxide in a process to develop SnO2 thin films, critical for electronics and optoelectronics, showcasing its utility in advanced material synthesis (VH Nguyen et al., 2022).

Features and Benefits

Precursor for OMCVD. Precursor for Sn-Al oxy alkoxide which may be heat-treated resulting in a zerovalent metal film. Ester elimination reactions lead to the rational synthesis of metal oxide materials.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service