Skip to Content
Merck
All Photos(1)

Key Documents

491799

Sigma-Aldrich

Aniline-1-13C

99 atom % 13C

Synonym(s):

Benzenamine-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC5H7N
CAS Number:
Molecular Weight:
94.12
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

refractive index

n20/D 1.586 (lit.)

bp

184 °C (lit.)

mp

−6 °C (lit.)

density

1.033 g/mL at 25 °C

mass shift

M+1

storage temp.

2-8°C

SMILES string

N[13c]1ccccc1

InChI

1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2/i6+1

InChI key

PAYRUJLWNCNPSJ-PTQBSOBMSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Resp. Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronald Besandre et al.
Organic letters, 15(7), 1666-1669 (2013-03-15)
N-substituted indoles are synthesized from primary amines through a tandem reaction sequence. Initial condensation of the amine with an α-(o-haloaryl)ketone or aldehyde is followed by intramolecular aryl amination catalyzed by CuI. A variety of anilines and alkyl amines, including those
Timo Stahl et al.
Journal of the American Chemical Society, 135(4), 1248-1251 (2013-01-15)
Heterolytic splitting of the Si-H bond mediated by a Ru-S bond forms a sulfur-stabilized silicon cation that is sufficiently electrophilic to abstract fluoride from CF(3) groups attached to selected anilines. The ability of the Ru-H complex, generated in the cooperative
Roman Spesyvtsev et al.
Faraday discussions, 157, 165-179 (2012-12-13)
Femtosecond time-resolved photoelectron imaging is employed to investigate ultrafast electronic relaxation in aniline, a prototypical aromatic amine. The molecule is excited at wavelengths between 269 and 238 nm. We observe that the S2(pi3s/pi sigma*) state is populated directly during the
Laya Siroos Rezaei et al.
Environmental technology, 33(19-21), 2273-2280 (2013-02-12)
Aerobic granules can be formed in sequencing batch airlift reactors (SBAR) and sequencing batch reactors (SBR). Comparing these two systems, the SBAR has excellent mixing condition, but due to a high height-to-diameter ratio (H/D), there is no performance capability at
Yu Qian et al.
Chemical communications (Cambridge, England), 49(26), 2700-2702 (2013-02-26)
An enantioselective four-component reaction of a diazoketone, water, an aniline and ethyl glyoxylate in the presence of catalytic Rh2(OAc)4 and a chiral Brønsted acid was developed to efficiently produce β-hydroxy-α-amino acid derivatives in good yields with high diastereoselectivity and enantioselectivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service