Skip to Content
Merck
All Photos(4)

Documents

490814

Sigma-Aldrich

Sodium nitrite-15N

98 atom % 15N, 95% (CP)

Synonym(s):

15N Labeled sodium nitrite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na15NO2
CAS Number:
Molecular Weight:
69.99
MDL number:
UNSPSC Code:
12352118
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

Quality Level

Assay

95% (CP)

form

solid

mp

271 °C (lit.)

mass shift

M+1

SMILES string

[Na+].[O-][15N]=O

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1/i1+1;

InChI key

LPXPTNMVRIOKMN-YTBWXGASSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Capilla Mata-Pérez et al.
Frontiers in plant science, 11, 962-962 (2020-07-28)
Nitro-fatty acids (NO2-FAs) are novel molecules resulting from the interaction of unsaturated fatty acids and nitric oxide (NO) or NO-related molecules. In plants, it has recently been described that NO2-FAs trigger an antioxidant and a defence response against stressful situations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service