Skip to Content
Merck
All Photos(1)

Key Documents

484164

Sigma-Aldrich

Carbon

glassy, spherical powder, 2-12 μm, 99.95% trace metals basis

Synonym(s):

Carbon

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99.95% trace metals basis

form

glassy, spherical powder

mol wt

Mw 12.011 g/mol

composition

C

particle size

2-12 μm

density

1.8-2.1 g/cm3

application(s)

battery manufacturing

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Glassy carbon shows only short range order in its structure. Photoacoustic doppler effect from the suspension of spherical carbon particles was investigated.[1] Glassy carbon exhibits a specific conductivity of 700Scm-1. 4

Application

Amorphous carbon may be used to develop zero temperature coefficient of resistance (TCR) based systems. [2] Glassy carbon has been used as a catalyst support for bimetallic Rh–ReOx. [3] It may be used to enhance the accuracy of infrared (IR) surface temperature measurements on micro-electro-mechanical systems (MEMS). [4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

From 5-hydroxymethylfurfural (HMF) to polymer precursors: catalyst screening studies on the conversion of 1, 2, 6-hexanetriol to 1, 6-hexanediol.
Buntara T, et al.
Topics in Catalysis, 55(5-7), 612-619 (2012)
Photoacoustic doppler effect from flowing small light-absorbing particles.
Fang H, et al.
Physical Review Letters, 99, 184501-184501 null
Smart conducting polymer composites having zero temperature coefficient of resistance
Chu K, et al.
Nanoscale, 7, 471-478 (2015)
Use of carbon microparticles for improved infrared temperature measurement of CMOS MEMS devices.
Hopper RH, et al.
Measurement Science and Technology, 21 null
I N Kurchenko et al.
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993), 75(3), 12-23 (2013-07-23)
The ability of soil and endophytic strains of P. funiculosum to accumulate biomass under cultivation conditions in media containing carbon sources from mono- to polysaccharides has been studied. It has been shown that the most favorable carbon sources for the

Articles

Recent demand for electric and hybrid vehicles, coupled with a reduction in prices, has caused lithium-ion batteries (LIBs) to become an increasingly popular form of rechargeable battery technology.

Solid oxide fuel cells and electrolyzers show potential for chemical-to-electrical energy conversion, despite early development stages.

Solid oxide fuel cells and electrolyzers show potential for chemical-to-electrical energy conversion, despite early development stages.

Solid oxide fuel cells and electrolyzers show potential for chemical-to-electrical energy conversion, despite early development stages.

See All

Questions

1–3 of 3 Questions  
  1. 비표면적(BET)가 얼마정도 되나요?

    1 answer
    1. The surface area for this product is not determined on a lot to lot basis. However, historical data indicates a surface area range of 1-2 m²/g. Please see below for additional physical properties of this material.
      Maximum service temperature (inert gas): 3000 °C
      Electrical resistance: 45 x 10-4 ?·cm
      Open porosity: 0 %
      Permeability coefficient:10-9 cm2/s
      Vickers hardness: HV1 230
      Coefficient of thermal expansion (20-200°C): 2.6 x 10-6 1/K
      Thermal conductivity: 6.3 W/(K x m)

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service