Skip to Content
Merck
All Photos(2)

Documents

479306

Sigma-Aldrich

cis-Diamineplatinum(II) dichloride

≥99.9% trace metals basis

Synonym(s):

cis-Diammineplatinum(II) dichloride, cis-Dichlorodiammine platinum(II), cis-Platinum(II) diammine dichloride, Cisplatin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pt(NH3)2Cl2
CAS Number:
Molecular Weight:
300.05
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.9% trace metals basis

reaction suitability

core: platinum
reagent type: catalyst

mp

270 °C (lit.)

antibiotic activity spectrum

neoplastics

Mode of action

DNA synthesis | interferes

SMILES string

N.N.Cl[Pt]Cl

InChI

1S/2ClH.2H3N.Pt/h2*1H;2*1H3;/q;;;;+2/p-2

InChI key

LXZZYRPGZAFOLE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

cis-Diamineplatinum(II) dichloride can be used as a model drug to test the efficiency of various drug carriers in drug delivery.
Highly active chemotherapeutic agent for use in preventing the advancement of testicular cancer.

Biochem/physiol Actions

Potent platinum-based antineoplastic agent. Forms cytotoxic adducts with the DNA dinucleotide d(pGpG), inducing intrastrand cross-links.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Carc. 1B - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly stable core-surface-crosslinked nanoparticles as cisplatin carriers for cancer chemotherapy.
Xu P, et al.
Colloids and Surfaces. B, Biointerfaces, 48(1), 50-57 (2006)
Synthesis of a new nanoparticle system based on electrostatic alginate?piperazine interactions.
Roman JV, et al.
Polymers For Advanced Technologies, 27(5), 623-629 (2016)
Pedro J Beltran et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(11), 2947-2958 (2014-04-15)
Insulin-like growth factor 1 receptor (IGF-IR) has been implicated in the pathogenesis of ovarian cancer. Ganitumab is an investigational, fully human monoclonal antibody against IGF-IR. Here, we explore the therapeutic potential of ganitumab for the treatment of ovarian cancer. The
Koji Tanaka et al.
Carcinogenesis, 36(8), 894-903 (2015-05-31)
There is increasing evidence that the expression of microRNA (miRNA) in cancer is associated with chemosensitivity but the mechanism of miRNA-induced chemoresistance has not been fully elucidated. The aim of this study was to examine the role of extracellular miRNA
H Sun et al.
British journal of cancer, 111(2), 281-291 (2014-06-20)
We investigated the biologic and pharmacologic activities of a chromosome region maintenance 1 (CRM1) inhibitor against human non-small cell lung cancer (NSCLC) cells both in vitro and in vivo. The in vitro and in vivo effects of a novel CRM1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service