Skip to Content
Merck
All Photos(1)

Documents

47718

Sigma-Aldrich

(Formylmethyl)triphenylphosphonium chloride

≥97.0% (AT)

Synonym(s):

2-Oxoethyltriphenylphosphonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOCH2P(Cl)(C6H5)3
CAS Number:
Molecular Weight:
340.78
Beilstein:
4060813
EC Number:
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (AT)

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

209-212 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Cl-].O=CC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C20H18OP.ClH/c21-16-17-22(18-10-4-1-5-11-18,19-12-6-2-7-13-19)20-14-8-3-9-15-20;/h1-16H,17H2;1H/q+1;/p-1

InChI key

RVEJRPJGKXTQIF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Hydroazidation reactions in preparation of a-azido alcohols
  • Preparation of prodrug (2-hydroxyamino-vinyl)-triphenyl-phosphonium (HVTP) inhibitor of peroxidase activity and apoptosis
  • Intramolecular [4 + 3] cycloaddition reactions

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jamal Bouitbir et al.
Toxicology, 426, 152281-152281 (2019-08-25)
Sunitinib is cardiotoxic, but the mechanisms are not entirely clear. We aimed to enlarge our knowledge about the role of mitochondria in cardiac toxicity of sunitinib in vitro and in vivo. For this reason, we studied the toxicity of sunitinib

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service