Skip to Content
Merck
All Photos(1)

Key Documents

474711

Sigma-Aldrich

Barium

dendritic pieces, purified by distillation, 99.99% trace metals basis

Synonym(s):

Barium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ba
CAS Number:
Molecular Weight:
137.33
EC Number:
MDL number:
UNSPSC Code:
12141505
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

dendritic pieces

purified by

distillation

reaction suitability

reagent type: reductant

resistivity

50.0 μΩ-cm, 20°C

bp

1640 °C (lit.)

mp

725 °C (lit.)

density

3.6 g/mL at 25 °C (lit.)

SMILES string

[Ba]

InChI

1S/Ba

InChI key

DSAJWYNOEDNPEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Barium is a reactive, soft electropositive metal with a body-centered cubic crystal structure. It can be produced in a vacuum by reducing its oxide with aluminum. It has medium-specific weight and good electrical conductivity. It is widely used in alloying, removing unwanted gases from vacuum tubes, and as a reducing agent.

Application

Barium can be used as a precursor to synthesize Ba3Bi6.67Se13,Ba3Bi6PbSe13, and Ba3Bi6SnSe13.

It can be used as a dopant for Pb2+replacement in perovskite films. The addition of barium enhances the power conversion efficiency of organic-inorganic lead halide solar cells.
Critical for the synthesis of pure Ba21M2O5HX (M = Ge, SI, Ga, In, Tl) suboxide Zintl phases.3

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang, C.H., et al.
Nanotech. in Catalysis, 2, 543-543 (2004)
Bryce J B Nelson et al.
Scientific reports, 10(1), 22203-22203 (2020-12-19)
This study reports the high-yield production of a novel 133/135La theranostic pair at a 22 MeV proton beam energy as an attractive alternative to the recently introduced 132/135La pair, demonstrating over an order of magnitude production increase of 133/135La (231 ± 8 MBq 133La
Haifeng Zheng et al.
American journal of physiology. Cell physiology, 306(7), C705-C713 (2014-01-31)
Interstitial cells of Cajal (ICC) generate slow waves in gastrointestinal (GI) muscles. Previous studies have suggested that slow wave generation and propagation depends on a voltage-dependent Ca(2+) entry mechanism with the signature of a T-type Ca(2+) conductance. We studied voltage-dependent
André Deutschmann et al.
Molecular pharmacology, 83(2), 501-511 (2012-12-01)
Bisphenol A (BPA), a high volume production chemical compound attracts growing attention as a health-relevant xenobiotic in humans. It can directly bind to hormone receptors, enzymes, and ion channels to become biologically active. In this study we show that BPA
Christine Austin et al.
Nature, 498(7453), 216-219 (2013-05-24)
Early-life dietary transitions reflect fundamental aspects of primate evolution and are important determinants of health in contemporary human populations. Weaning is critical to developmental and reproductive rates; early weaning can have detrimental health effects but enables shorter inter-birth intervals, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service