All Photos(2)
About This Item
Linear Formula:
C5H11NH2
Molecular Weight:
87.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥98%
refractive index
n20/D 1.411 (lit.)
bp
104 °C (lit.)
mp
−55 °C (lit.)
density
0.752 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CCCCCN
InChI
1S/C5H13N/c1-2-3-4-5-6/h2-6H2,1H3
InChI key
DPBLXKKOBLCELK-UHFFFAOYSA-N
Related Categories
General description
The product is a mixture of isomeric amylamines. Amylamine is a simple primary amine. Neopentylamine and tert-amylamine are its isomeric forms. The mechanism of the hydrodenitrogenation of these isomeric amylamines has been studied in the presence of sulfided NiMo/A12O3 catalyst. Kinetics and thermodynamic parameters of the ultrasonic absorptions of its aqueous solutions have been evaluated.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
42.8 °F
Flash Point(C)
6 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mechanism of carbon-nitrogen bond cleavage during amylamine hydrodenitrogenation over a sulphided NiMo/Al2O3 catalyst.
Portefaix JL, et al.
Catalysis Letters, 9(1-2), 127-132 (1991)
Kinetic Studies of Fast Reactions in Aqueous Solutions of Amylamine by Means of Ultrasonic Absorption.
Nishikawa S, et al.
Bulletin of the Chemical Society of Japan, 46(10), 2992-2997 (1973)
Jun Kobayashi et al.
Journal of biomedical materials research. Part A, 102(11), 3883-3893 (2013-12-18)
Antibody-immobilized thermoresponsive poly(N-isopropylacrylamide-co-2-carboxyisopropylacrylamide) [poly(IPAAm-co-CIPAAm)]-grafted cell culture surfaces were designed to enhance both the initial adhesion of weakly adhering cells and the ability of cells to detach in response to low temperature through the regulation of affinity binding between immobilized antibodies
Cui Cui et al.
Bone, 72, 43-52 (2014-12-03)
Serotonin (5-HT)--a monoamine with a variety of physiological functions--has recently emerged as a major regulator of bone mass. 5-HT is synthesized in the brain and the gut, and gut-derived 5-HT contributes to circulating 5-HT levels and is a negative modulator
Renata Grzywa et al.
Chembiochem : a European journal of chemical biology, 15(17), 2605-2612 (2014-09-23)
Neutrophils are a type of granulocyte important in the "first line of defense" of the innate immune system. Upon activation, they facilitate the destruction of invading microorganisms by the production of superoxide radicals, as well as the release of the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service