Skip to Content
Merck
All Photos(2)

Key Documents

47299

Sigma-Aldrich

5-(Fmoc-amino)-1-pentanol

≥98.0% (HPLC)

Synonym(s):

9-Fluorenylmethyl N-(5-hydroxypentyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23NO3
Molecular Weight:
325.40
Beilstein:
3623714
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥98.0% (HPLC)

Quality Level

Assay

≥98.0% (HPLC)

reaction suitability

reagent type: cross-linking reagent

mp

103-108 °C (dec.)

functional group

Fmoc
hydroxyl

storage temp.

2-8°C

SMILES string

OCCCCCNC(OCC1C(C=CC=C2)=C2C3=C1C=CC=C3)=O

InChI

1S/C20H23NO3/c22-13-7-1-6-12-21-20(23)24-14-19-17-10-4-2-8-15(17)16-9-3-5-11-18(16)19/h2-5,8-11,19,22H,1,6-7,12-14H2,(H,21,23)

InChI key

YNOWFUNORLDFTH-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service