Skip to Content
Merck
All Photos(3)

Key Documents

471593

Sigma-Aldrich

N,N-Diethylhydroxylamine

≥98%

Synonym(s):

N-Hydroxydiethylamine, DEHA, Diethylhydroxylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2NOH
CAS Number:
Molecular Weight:
89.14
Beilstein:
1731349
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.07 (vs air)

Quality Level

vapor pressure

4 mmHg ( 0 °C)

Assay

≥98%

expl. lim.

10 %

refractive index

n20/D 1.420 (lit.)

bp

125-130 °C (lit.)

mp

−26-−25 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCN(O)CC

InChI

1S/C4H11NO/c1-3-5(6)4-2/h6H,3-4H2,1-2H3

InChI key

FVCOIAYSJZGECG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Diethylhydroxylamine (diethylhydroxylamine, DEHA, (C2H5)2NOH) is a N,N-dialkylhydroxylamine. Its interaction with the surface of clean silicon(100)-2x1 has been studied by various theoretical and spectroscopic methods. H2O2 along with superoxide and diethylnitroxide radicals have been reported to be formed during the photochemical oxidation of DEHA.

Application

N,N-Diethylhydroxylamine may be employed as a ligand in the preparation of unsymmetric mixed ligand oxadiazoline and/or imine platinum complexes. It may be used in the synthesis of organometallic clusters of mixed hydrazide/hydroxylamide clusters of zinc.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jamal Lasri et al.
Dalton transactions (Cambridge, England : 2003), (30)(30), 3259-3266 (2007-09-26)
Iminoacylation of acetone oxime Me(2)C[double bond, length as m-dash]NOH upon reaction with trans-[PtCl(2)(NCCH(2)CO(2)Me)(2)] and [2 + 3] cycloaddition of acyclic nitrone (-)O(+)N(Me) = C(H)(C(6)H(4)Me-4) to a nitrile ligand in lead to the formation of mono-imine trans-[PtCl(2)(imine-a)(NCCH(2)CO(2)Me)] [imine-a = NH[double bond
Hyunsung Mun et al.
Polymers, 11(6) (2019-06-15)
In the tire industry, solution styrene butadiene rubber (SSBR), which can introduce a functional group with good reactivity to silica at chain ends, is used to increase rolling resistance performance by considering fuel economy. However, this is not environmentally friendly
E Kurisaki et al.
The American journal of forensic medicine and pathology, 12(1), 54-58 (1991-03-01)
A 39-year-old man committed suicide by ingesting a large quantity of SH792. SH792 is a silicone resin precursor used as a hardener for waterproof paints. It is polymerized in water; this process is then followed by the formation of silicone
Gino A DiLabio et al.
Physical chemistry chemical physics : PCCP, 9(13), 1629-1634 (2007-04-13)
Incorporating diversity into structures constructed from the organic modification of silicon surfaces requires the use of molecules that contain multiple substituents of different types. In this work we examine the possible dissociation pathways of diethylhydroxylamine (DEHA, (C(2)H(5))(2)NOH) on the surface
P Bilski et al.
Photochemistry and photobiology, 58(1), 11-18 (1993-07-01)
The photooxidation of N,N-diethylhydroxylamine (DEHA) by Rose Bengal (RB) has been investigated in micellar and nonmicellar aqueous solutions. We measured the quantum yield of oxygen consumption forming H2O2 and monitored two intermediates, the superoxide and diethylnitroxide radicals. When the pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service