Skip to Content
Merck
All Photos(1)

Key Documents

467731

Sigma-Aldrich

Ammonium bromide

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Br
CAS Number:
Molecular Weight:
97.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 198.3 °C)

Assay

≥99.99% trace metals basis

form

powder and chunks

impurities

≤0.005% insolubles
<100 ppm total metallic impurities

ign. residue

≤0.01%

pH

4.5-6.0

bp

396 °C/1 atm (lit.)

mp

452 °C (lit.)

solubility

acetone: soluble(lit.)
diethyl ether: slightly soluble(lit.)
ethanol: soluble(lit.)

density

2.43 g/mL at 25 °C (lit.)

anion traces

bromate (BrO3-): ≤0.002%
chloride (Cl-): ≤0.2%
iodide (I-): ≤0.005%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.002%
Fe: ≤5 ppm
heavy metals: ≤5 ppm

SMILES string

N.Br[H]

InChI

1S/BrH.H3N/h1H;1H3

InChI key

SWLVFNYSXGMGBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Lact. - Repr. 1B - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emilia Fisicaro et al.
The journal of physical chemistry. B, 109(5), 1744-1749 (2006-07-21)
The apparent and partial molar enthalpies, apparent molar volumes, and adiabatic compressibilities at 298 K of the aqueous solutions of the cationic gemini surfactants propanediyl-alpha,omega-bis(octyldimethylammonium bromide) (8-3-8) and propanediyl-alpha,omega-bis(dodecyldimethylammonium bromide) (12-3-12) have been measured as a function of concentration. The
Dong Xie et al.
Dental materials : official publication of the Academy of Dental Materials, 27(5), 487-496 (2011-03-11)
The objective of this study was to use the newly synthesized poly(quaternary ammonium salt) (PQAS)-containing polyacid to formulate the light-curable glass-ionomer cements and study the effect of the PQAS on the compressive strength and antibacterial activity of the formed cements.
Gulnara A Gainanova et al.
Journal of colloid and interface science, 367(1), 327-336 (2011-12-03)
A systematic study of the aggregation behavior of alkyltriphenylphosphonium bromides (TPPB-n; n=8, 10, 12, 14, 16, 18; here n is the number of carbon atoms in alkyl groups) in aqueous solutions has been carried out and compared with trimethyl ammonium
Qin Zhou et al.
Journal of colloid and interface science, 314(2), 405-414 (2007-08-04)
Water purification is of extreme importance worldwide. p-Nitrophenol was used as a test chemical to design and test an organoclay for the removal of p-nitrophenol from an aqueous solution. Synthesis of the organoclay with methyltrioctadecylammonium bromide [CH(3)(CH(2))(17)](3)NBr(CH(3)) labeled as MTOAB
João A S Almeida et al.
Physical chemistry chemical physics : PCCP, 12(43), 14462-14476 (2010-10-05)
Gemini surfactants possess interesting interfacial and aggregation properties that have prompted comprehensive studies and successful applications in a wide variety of fields. However, a systematic study on the effect of gemini tail and spacer length upon the organization of lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service