Skip to Content
Merck
All Photos(4)

Documents

461350

Sigma-Aldrich

1-Boc-4-piperidone

98%

Synonym(s):

N-Boc-4-piperidone, tert-Butyl 4-oxo-1-piperidinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO3
CAS Number:
Molecular Weight:
199.25
Beilstein:
3650236
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

73-77 °C (lit.)

SMILES string

CC(C)(C)OC(=O)N1CCC(=O)CC1

InChI

1S/C10H17NO3/c1-10(2,3)14-9(13)11-6-4-8(12)5-7-11/h4-7H2,1-3H3

InChI key

ROUYFJUVMYHXFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Boc-4-piperidone is used as a precursor in the synthesis of synthetic drugs and as a building block for the preparation of selective ligands.

Application

1-Boc-4-piperidone, a pharma building block, may be used in the synthesis of (3E,5E)-3,5-bis(2,5-dimethoxybenzylidene)-1-t-butoxycarbonylpiperidin-4-one (RL197). It may also be used in the synthesis of spirorifamycins containing a piperidine ring structure.

Warning

Avoid metal contact.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Design, synthesis, and structure--activity relationships of novel spiro-piperidines as acetyl-CoA carboxylase inhibitors
Kamata, Makoto and Yamashita
Bioorganic & Medicinal Chemistry Letters, 22, 3643-3647 (2012)
John W Clader et al.
Bioorganic & medicinal chemistry, 12(2), 319-326 (2004-01-16)
Anthranilamide analogues such as 23 are potent and highly selective muscarinic M2 antagonists that also show good oral bioavailability and in vivo activity.
Gebhard Thoma et al.
Journal of medicinal chemistry, 47(8), 1939-1955 (2004-04-02)
The chemokine receptor CCR5 plays an important role in inflammatory and autoimmune disorders as well as in transplant rejection by affecting the trafficking of effector T cells and monocytes to diseased tissues. Antagonists of CCR5 are believed to be of
The evolving regulatory landscape for fentanyl: China, India, and global drug governance
Wang, Chao and Lassi
International Journal of Environmental Research and Public Health, 19, 2074-2074 (2022)
Atsuya Takami et al.
Bioorganic & medicinal chemistry, 12(9), 2115-2137 (2004-04-15)
Several structurally unrelated scaffolds of the Rho kinase inhibitor were designed using pharmacophore information obtained from the results of a high-throughput screening and structural information from a homology model of Rho kinase. A docking simulation using the ligand-binding pocket of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service