Skip to Content
Merck
All Photos(1)

Key Documents

460273

Sigma-Aldrich

1,2-Bis(diphenylphosphino)benzene

97%

Synonym(s):

1,2-Bis(diphenylphosphanyl)benzene, dppBz, dppbe, dppben, dppbenz, dppbz, o-Phenylenebis[diphenylphosphine], o-bis(diphenylphosphino)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4[P(C6H5)2]2
CAS Number:
Molecular Weight:
446.46
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: ligand
reaction type: Baeyer-Villiger Oxidation

reagent type: ligand
reaction type: Cycloadditions

reagent type: ligand
reaction type: Reductions

mp

183-188 °C (lit.)

functional group

phosphine

SMILES string

c1ccc(cc1)P(c2ccccc2)c3ccccc3P(c4ccccc4)c5ccccc5

InChI

1S/C30H24P2/c1-5-15-25(16-6-1)31(26-17-7-2-8-18-26)29-23-13-14-24-30(29)32(27-19-9-3-10-20-27)28-21-11-4-12-22-28/h1-24H

InChI key

NFRYVRNCDXULEX-UHFFFAOYSA-N

Related Categories

Application

1,2-Bis(diphenylphosphino)benzene (DPPB) can be used:
  • As a chelating ligand in the synthesis of luminescent copper(I) halide complexes.
  • In the synthesis of copper-1,2-bis(diphenylphosphino)benzene catalyst which is used for the β-boration of α, β-unsaturated amide.
  • As an alternative to TMEDA for the selective cleavage of sp3 C-X bond in fluoroaromatic coupling reactions catalyzed by iron.
  • As a ligand in the synthesis of alkenylboronates from acetylenic esters.
  • To synthesize copper-diphosphine complexes which are used as homogeneous catalysts for N-formylation of a wide range of amines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-Catalyzed Conjugate Addition of Diboron Reagents to α, β -Unsaturated Amides: Highly Reactive Copper-1, 2-Bis (diphenylphosphino) benzene Catalyst System
Chea H, et al.
advanced synthesis and catalysis, 351(6), 855-858 (2009)
Iron-catalysed fluoroaromatic coupling reactions under catalytic modulation with 1, 2-bis (diphenylphosphino) benzene
Hatakeyama T, et al.
Chemical Communications (Cambridge, England), 351(10), 1216-1218 (2009)
Photophysical properties of highly luminescent copper (I) halide complexes chelated with 1, 2-bis (diphenylphosphino) benzene
Tsuboyama A, et al.
Inorganic Chemistry, 46(6), 1992-2001 (2007)
Synthesis of Activated Alkenylboronates from Acetylenic Esters by CuH-Catalyzed 1, 2-Addition/Transmetalation
Lipshutz BH, et al.
Angewandte Chemie (International Edition in English), 47(52), 10183-10186 (2008)
Aparajita Mukherjee et al.
Dalton transactions (Cambridge, England : 2003), 47(30), 10264-10272 (2018-07-18)
Reaction of 1,2-bis(diphenylphosphino)benzene (dppbz) with [{Ru(CO)2Cl2}n] affords [Ru(dppbz)(CO)2Cl2], where the two carbonyls are mutually cis and the two chlorides are trans. The molecular structure of [Ru(dppbz)(CO)2Cl2], has been determined by X-ray crystallography, and the stability of the different available stereoisomers

Articles

The Baeyer-Villiger oxidation is the oxidative cleavage of a carbon-carbon bond adjacent to a carbonyl, which converts the ketones to esters and the cyclic ketones to lactones.

The Baeyer-Villiger oxidation is the oxidative cleavage of a carbon-carbon bond adjacent to a carbonyl, which converts the ketones to esters and the cyclic ketones to lactones.

The Baeyer-Villiger oxidation is the oxidative cleavage of a carbon-carbon bond adjacent to a carbonyl, which converts the ketones to esters and the cyclic ketones to lactones.

The Baeyer-Villiger oxidation is the oxidative cleavage of a carbon-carbon bond adjacent to a carbonyl, which converts the ketones to esters and the cyclic ketones to lactones.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service