Skip to Content
Merck
All Photos(1)

Key Documents

455822

Sigma-Aldrich

Ammonium perchlorate

99.999% trace metals basis

Synonym(s):

Ammonium perchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4ClO4
CAS Number:
Molecular Weight:
117.49
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

solid

reaction suitability

reagent type: oxidant

impurities

≤20.0 ppm Trace Metal Analysis

density

1.95 g/mL at 25 °C (lit.)

SMILES string

N.OCl(=O)(=O)=O

InChI

1S/ClHO4.H3N/c2-1(3,4)5;/h(H,2,3,4,5);1H3

InChI key

HHEFNVCDPLQQTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 1 - STOT RE 2

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phillip E Morgan et al.
Biomedical chromatography : BMC, 24(3), 318-323 (2009-07-28)
HPLC columns packed with 3 microm particle size HPLC Technology Techsphere SCX (propylsulfonic acid-modified) silica offer considerable advantages over 5 microm SCX packings in the analysis of basic drugs using 100% methanol eluents containing an ionic modifier such as ammonium
T E Stoker et al.
Toxicology, 228(1), 58-65 (2006-10-03)
The U.S. EPA Endocrine Disruptor Screening Program (EDSP) Tier 1 male pubertal protocol was designed as a screen to detect endocrine-disrupting chemicals which may alter reproductive development or thyroid function. One purpose of this in vivo screening protocol is to
Qi Yang et al.
Journal of hazardous materials, 197, 199-203 (2011-10-18)
An energetic coordination compound Cu(Mtta)(2)(NO(3))(2) has been synthesized by using 1-methyltetrazole (Mtta) as ligand and its structure has been characterized by X-ray single crystal diffraction. The central copper (II) cation was coordinated by four O atoms from two Mtta ligands
Geneviève Bordeleau et al.
Journal of environmental quality, 37(2), 308-317 (2008-02-13)
Within Canada, it has been recognized in the last decade that military training activities may have impacts on the environmental quality of training ranges. However, impacts of activities specific to Air Force Bases have not yet been intensely documented. A
Yu Chen et al.
General and comparative endocrinology, 159(2-3), 196-207 (2008-09-23)
Perchlorate, a known thyroid disruptor, is deposited in eggs of exposed female birds, raising concerns that the embryos from these eggs may become hypothyroid, which may in turn affect the development and function of thyroid-dependent organs. We hypothesized that exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service