Skip to Content
Merck
All Photos(1)

Key Documents

449954

Sigma-Aldrich

Neodymium(III) fluoride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Neodymium fluoride, Neodymium trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NdF3
CAS Number:
Molecular Weight:
201.24
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: neodymium

impurities

≤150.0 ppm Trace Rare Earth Analysis

bp

2300 °C (lit.)

density

6.65 g/mL at 25 °C (lit.)

SMILES string

F[Nd](F)F

InChI

1S/3FH.Nd/h3*1H;/q;;;+3/p-3

InChI key

XRADHEAKQRNYQQ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Used extensively in the preparation and study of fluorinated glasses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Journal of Materials Science Letters, 15, 1854-1854 (1996)
Journal of Non-Crystalline Solids, 204, 178-178 (1996)
Z S Žunic et al.
Radiation protection dosimetry, 184(3-4), 319-323 (2019-04-30)
Urine, hair and nail samples were collected from a population of both sex aged from 40 to 87 years in Niška Banja (227 km south of Belgrade, Serbia) with a high level of natural radioactivity (HLNRA). To assess and monitor
J. Alloy Compounds, 232, 53-53 (1996)
Ferroelectr., Lett. Sect., 21, 111-111 (1996)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service