Skip to Content
Merck
All Photos(3)

Key Documents

448877

Sigma-Aldrich

Chitosan

Chitosan

medium molecular weight

Synonym(s):

Deacetylated chitin, Poly(D-glucosamine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder

viscosity

200-800 cP, 1 wt. % in 1% acetic acid(25 °C, Brookfield)(lit.)

solubility

dilute aqueous acid: soluble

SMILES string

N([C@H]1[C@@H](O[C@@H]([C@H]([C@@H]1O)O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4N)O)O[C@@H]5O[C@@H]([C@H]([C@@H]([C@H]5N)O)O[C@@H]6O[C@@H]([C@H]([C@@H]([C@H]6N)O)O[C@@H]7O[C@@H]([C@H]([C@@H]([C@H]7N)O)O[C@@H]8O[C@@H]([C@H]([C@@H]([C@H]8N)O)O[C@@H]9O[C@@H]([C@H](

InChI

1S/C56H103N9O39/c1-87-56(86)65-28-38(84)46(19(10-74)96-55(28)104-45-18(9-73)95-49(27(64)37(45)83)97-39-12(3-67)88-47(85)20(57)31(39)77)103-54-26(63)36(82)44(17(8-72)94-54)102-53-25(62)35(81)43(16(7-71)93-53)101-52-24(61)34(80)42(15(6-70)92-52)100-51-23(60)33(79)41(14(5-69)91-51)99-50-22(59)32(78)40(13(4-68)90-50)98-48-21(58)30(76)29(75)11(2-66)89-48/h11-55,66-85H,2-10,57-64H2,1H3,(H,65,86)/t11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48+,49+,50+,51+,52+,53+,54+,55+/m1/s1

InChI key

FLASNYPZGWUPSU-SICDJOISSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chitosan may be used as a flocculant, in protein precipitation, encapsulating agent and aqueous thickener.
Forms gels with multivalent anions. Gives clear solutions that dry to strong, clear films.

Features and Benefits

Biocompatible, antibacterial and environmentally friendly polyelectrolyte with a variety of applications including water treatment, chromatography, additives for cosmetics, textile treatment for antimicrobial activity, novel fibers for textiles, photographic papers, biodegradable films, biomedical devices, and microcapsule implants for controlled release in drug delivery.
Biocompatible, antibacterial and environmentally friendly polyelectrolyte.

Physical form

75-85% deacetylated

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arnout Mieremet et al.
PloS one, 12(3), e0174478-e0174478 (2017-03-24)
Full thickness human skin models (FTMs) contain an epidermal and a dermal equivalent. The latter is composed of a collagen dermal matrix which harbours fibroblasts. Current epidermal barrier properties of FTMs do not fully resemble that of native human skin
Viviana Vergaro et al.
International journal of biological macromolecules, 99, 187-195 (2017-02-25)
Carbohydrate polimeric microcapsules were assembled using a LbL approach onto a CaCO
Mohammad Amin Shamekhi et al.
International journal of biological macromolecules, 127, 396-405 (2019-01-10)
Enhancement of physical and mechanical properties of the scaffolds is achieved via various methods including cross-linking and incorporation of nano particles. In the present research chitosan-based scaffolds firstly were reinforced with the incorporation of the graphene oxide (GO) nanoparticles. The
Paulo V O Toledo et al.
International journal of biological macromolecules, 123, 1180-1188 (2018-11-24)
Xanthan gum (XG) was applied for the creation of hydrophilic, hydrophobic and layered cryogels. Firstly, the correlation among synthesis parameters, such as solvent composition and polymer concentration (Cp) in the precursor gel and mold diameter (Φ), with physicochemical properties and
Lana Glerieide Silva Garcia et al.
Carbohydrate polymers, 195, 662-669 (2018-05-29)
Difficulties in the treatment of Candida spp. invasive infections are usually related to the formation of biofilms. The aim of this study was to determine the effects of molecular weight (MW) of chitosan (using high (HMW), medium (MMW) and low

Articles

Carcinogenic compounds in elastomers: 1,3-butadiene and isoprene used in elastomer manufacturing, classified as carcinogenic by IARC.

Carcinogenic compounds in elastomers: 1,3-butadiene and isoprene used in elastomer manufacturing, classified as carcinogenic by IARC.

Polymers, natural or synthetic, are vital in medical advancements, contributing to device and pharmaceutical development in therapy.

Polymers, natural or synthetic, are vital in medical advancements, contributing to device and pharmaceutical development in therapy.

See All

Questions

1–10 of 15 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the particle size of this product (448877 chitosan)?

    1 answer
    1. The particle size of this product is not determined.

      Helpful?

  4. Is the chitosan produced starting from shrimp shells that are sourced as waste from the food industry, or are these shrimps specifically farmed for chitosan production?

    1 answer
    1. The shrimp sourced to produce this item are harvested in Iceland specifically for use in chitosan production.

      Helpful?

  5. What is the purity of chitosan?

    1 answer
    1. The minimum purity of this product is 75%. Purity is determined on a lot-specific basis.

      Helpful?

  6. Could you please provide us the chemical formula of this chitosan compound?

    1 answer
    1. Chitasan is a biopolymer with a base molecular formula of C12H24N2O9. This particular product is a medium molecular weight option with a range of 190,000-310,000 Da.

      Helpful?

  7. Do you perform a chemical or enzymatic deacetylation on Chitin to obtain Chitosan? Which chemical or enzyme do you employ?

    1 answer
    1. Unfortunately, the deacetylation method is considered proprietary.

      Helpful?

  8. Which Chitosan has the highest quality, for research or for cosmetic purposes?

    1 answer
    1. Unfortunately, none of the Chitosan products currently offered are suitable for cosmetic purposes.

      Helpful?

  9. What is the polydispersity of medium molecular weight chitosan, product 448877?

    1 answer
    1. Unfortunately, the polydispersity is not determined for for this item.

      Helpful?

  10. This medium molecular chitosan is from crab or shrimp? Or other way to produce?

    1 answer
    1. This chitosan is from Pandalus Borealis (shrimp).
      Please see the sample Certificate of Origin for more information: https://www.sigmaaldrich.com/coo/undefined/448877/*

      Helpful?

1–10 of 15 Questions  

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service