Skip to Content
Merck
All Photos(1)

Key Documents

448079

Sigma-Aldrich

3-Bromobenzyl chloride

98%

Synonym(s):

3-Bromo-α-chlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH2Cl
CAS Number:
Molecular Weight:
205.48
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.583 (lit.)

bp

116-118 °C/17 mmHg (lit.)

density

1.608 g/mL at 25 °C (lit.)

functional group

bromo
chloro

SMILES string

ClCc1cccc(Br)c1

InChI

1S/C7H6BrCl/c8-7-3-1-2-6(4-7)5-9/h1-4H,5H2

InChI key

UDKGXKYEWBGQCG-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile Chlorination of Benzyl Alcohols Using 1, 8-Diazabicyclo [5.4. 0] undec-7-ene (DBU) and Sulfonyl Chlorides.
Kim HW, et al.
ChemInform, 42(14) (2011)
Bromo Derivatives of Benzylphenols. I. Some Monobromo, Dibromo and Tribromo Derivatives of Ortho and Para Benzylphenols.
Huston RC, et al.
Journal of the American Chemical Society, 55(5), 2146-2149 (1933)
Y Magata et al.
Nuclear medicine and biology, 27(2), 163-168 (2000-04-25)
Use of the [(18)F]-fluoromethyl phenyl group is an attractive alternative to direct fluorination of phenyl groups because the fluorination of the methyl group takes place under milder reaction conditions. However, we have found that 4-FMeBWAY showed femur uptake equal to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service