Skip to Content
Merck
All Photos(2)

Key Documents

447218

Sigma-Aldrich

Trimethyl borate

99.999% (trace metal basis)

Synonym(s):

Boric acid trimethyl ester, Methyl borate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
B(OCH3)3
CAS Number:
Molecular Weight:
103.91
Beilstein:
1697939
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.59 (vs air)

Quality Level

Assay

99.999% (trace metal basis)

refractive index

n20/D 1.346 (lit.)

bp

68-69 °C (lit.)

mp

−34 °C (lit.)

density

0.932 g/mL at 20 °C (lit.)

SMILES string

COB(OC)OC

InChI

1S/C3H9BO3/c1-5-4(6-2)7-3/h1-3H3

InChI key

WRECIMRULFAWHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - (own results)

Flash Point(C)

-11 °C - (own results)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Colorado et al.
Journal of mass spectrometry : JMS, 31(4), 403-410 (1996-04-01)
Barbiturates are a class of drugs that are utilized as anesthetics and sleeping agents and are used for the treatment of anxiety, epilepsy and other psychiatric disorders. Because of their pyrimidine structures, barbiturates are highly basic compounds. The evaluation of
Jayalakshmi Somuramasami et al.
Journal of the American Society for Mass Spectrometry, 22(6), 1040-1051 (2011-09-29)
Several lignin model compounds were examined to test whether gas-phase ion-molecule reactions of trimethylborate (TMB) in a FTICR can be used to differentiate the ortho-, meta-, and para-isomers of protonated aromatic compounds, such as those formed during degradation of lignin.
R Meder et al.
Solid state nuclear magnetic resonance, 15(1), 69-72 (2000-07-21)
Boron-11 nuclear magnetic resonance imaging and spectroscopy have been used to characterise the nature and distribution of boron compounds after preservative treatment of radiata pine wood with trimethylborate (TMB). One day after treatment, 11B magnetic resonance imaging microscopy showed significant
Steven C Habicht et al.
Analytical chemistry, 80(9), 3416-3421 (2008-03-28)
A mass spectrometric method has been developed for the identification of the carboxylic acid functional group in analytes evaporated and ionized by electrospray ionization (ESI). This method is based on gas-phase ion-molecule reactions of ammoniated ([M + NH4]+) and sodiated
Scott Gronert et al.
Journal of the American Society for Mass Spectrometry, 13(9), 1088-1098 (2002-09-27)
Using a quadrupole ion trap mass spectrometer, trimethyl borate was allowed to react with dihydrogen phosphate, deprotonated O-phosphoserine, and a set of hydrogen bonded complexes involving dihydrogen phosphate and neutral acids (phosphoric acid, acetic acid, serine, and O-phosphoserine). The reactions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service