Skip to Content
Merck
All Photos(1)

Key Documents

445177

Sigma-Aldrich

O,O-Diethyl thiophosphate potassium salt

98%

Synonym(s):

Potassium O,O-diethyl phosphorothioate, Potassium O,O-diethylthiophosphate, Potassium di-O-ethyl phosphorothioate, Potassium diethyl thiophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2P(O)SK
CAS Number:
Molecular Weight:
208.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

180-186 °C (lit.)

SMILES string

[K+].CCOP([O-])(=S)OCC

InChI

1S/C4H11O3PS.K/c1-3-6-8(5,9)7-4-2;/h3-4H2,1-2H3,(H,5,9);/q;+1/p-1

InChI key

OVJCAYMARFNMQB-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Koutroulakis et al.
Reproductive toxicology (Elmsford, N.Y.), 46, 98-105 (2014-04-02)
The aim of this study was to evaluate fetal exposure to organophosphate pesticides (OPs) by measuring their non-specific dialkyl-phosphate metabolites (DAPs) in amniotic fluid (AF), and to examine the potential association between prenatal exposure and fetal growth. AF samples were
M E Mount
American journal of veterinary research, 45(4), 817-824 (1984-04-01)
Recognition of exposure to diazinon, an organophosphate insecticide, was studied in goats. Urine and milk dialkyl phosphate concentrations (DETP; O,O-diethyl phosphorothionate) and blood cholinesterase activity (ChE) and diazinon concentrations were measured. Groups (n = 3 each) given (orally) diazinon at
Jialong Jie et al.
Nucleic acids research, 47(22), 11514-11526 (2019-11-15)
Phosphorothioate (PS) modifications naturally appear in bacteria and archaea genome and are widely used as antisense strategy in gene therapy. But the chemical effects of PS introduction as a redox active site into DNA (S-DNA) is still poorly understood. Herein
Personnel exposure to diazinon in a supervised pest eradication program.
C P Weisskopf et al.
Archives of environmental contamination and toxicology, 17(2), 201-212 (1988-03-01)
J Fukal et al.
Physical chemistry chemical physics : PCCP, 21(19), 9924-9934 (2019-05-01)
Structural interpretation of the 31P NMR shifts measured in O,O-diethyl thiophosphate (PT), 5,5-dimethyl-2-mercapto-1,3,2-dioxaphosphorinane 2-oxide (cPT), diethylphosphate (P) and 5,5-dimethyl-2-hydroxy-1,3,2-dioxaphosphinane 2-oxide (cP) was obtained by means of theoretical calculations including the effects of geometry, molecular dynamics, and solvent, relativistic effects and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service