Skip to Content
Merck
All Photos(1)

Key Documents

444553

Sigma-Aldrich

2-Hydroxy-3-methylbenzaldehyde

98%

Synonym(s):

3-Methylsalicylaldehyde, 2,3-Cresotaldehyde, 2-Hydroxy-3-methylbenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(CH3)CHO
CAS Number:
Molecular Weight:
136.15
Beilstein:
1099377
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.564 (lit.)

bp

204 °C/730 mmHg (lit.)

density

1.123 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

Cc1cccc(C=O)c1O

InChI

1S/C8H8O2/c1-6-3-2-4-7(5-9)8(6)10/h2-5,10H,1H3

InChI key

IPPQNXSAJZOTJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[2-(1H-Benzimidazol-2-yl-?N3) aniline-?N] dichloridozinc.
Eltayeb NE, et al.
Acta Crystallographica Section E, Structure Reports Online, 67(8), 1062-1063 (2011)
Irina A Kühne et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 32(40), 404002-404002 (2020-03-26)
Structural, magnetic and electromechanical changes resulting from spin crossover between the spin quintet and spin triplet forms of a mononuclear Mn3+ complex embedded in six lattices with different charge balancing counterions are reported. Isostructural ClO4- and BF4- salts (1) and
8-Substituted 3-Arylcoumarins as Potent and Selective MAO-B Inhibitors: Synthesis, Pharmacological Evaluation, and Docking Studies.
Vi?a D, et al.
ChemMedChem, 7(3), 464-470 (2012)
Synthesis and characterization of amino acid Schiff base complexes of nickel (II).
Wang G and Chang JC.
Synth. React. Inorg. Met.-Org. Chem. , 24(7), 1091-1097 (1994)
P Valenti et al.
Bioorganic & medicinal chemistry, 6(6), 803-810 (1998-07-29)
A series of 1,4-dihydropyridines bearing a coumarin moiety in 4-position was synthesized. The compounds were evaluated for inotropic, chronotropic and calcium antagonist activities. The replacement of the o-nitrophenyl moiety of nifedipine with a coumarin or phenylcoumarin system is accompanied by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service