Skip to Content
Merck
All Photos(2)

Key Documents

444413

Sigma-Aldrich

N-(tert-Butoxycarbonyl)-L-valinol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH[NHCO2C(CH3)3]CH2OH
CAS Number:
Molecular Weight:
203.28
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

optical activity

[α]23/D −23°, c = 1 in chloroform

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.449 (lit.)

bp

208 °C (lit.)

density

0.995 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)[C@@H](CO)NC(=O)OC(C)(C)C

InChI

1S/C10H21NO3/c1-7(2)8(6-12)11-9(13)14-10(3,4)5/h7-8,12H,6H2,1-5H3,(H,11,13)/t8-/m1/s1

InChI key

OOQRRYDVICNJGC-MRVPVSSYSA-N

Application

Starting material for the synthesis of enantiopure homo-β-amino acids. Used in the efficient synthesis of enantiopure tetrahydroisoquinolines. Intermediate in the one-pot conversion of amino acid carbamates to N-derivatized 2-oxazolidinones.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huwe, C.M. Blechert, S.
Tetrahedron Letters, 35, 9533-9533 (1994)
Caputo, R. et al.
Tetrahedron, 51, 12337-12337 (1995)
R. Caputo et al.
Tetrahedron Letters, 36, 167-167 (1995)
Tietze, L.F. Burkhardt, O.
Synthesis, 1331-1331 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service